1-(2-Aminoethyl)-1H-1,2,3-triazole-4-carboxylic acid hydrochloride ≥90%
Supplier: Matrix Scientific
115130-1G
115130-500MG
102895-468EA
693.85
USD
102895-468
102895-470
1-(2-Aminoethyl)-1H-1,2,3-triazole-4-carboxylic acid hydrochloride ≥90%
1-(2-Aminoethyl)-1H-1,2,3-triazole-4-carboxylic acid hydrochloride
Hazard: Irritant
Formula:
C₅H₉ClN₄O₂ |
MDL Number:
MFCD28023526 CAS Number: 1232979-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...