4-(6,8-Dioxo-5,8-dihydro[1,3]dioxolo[4,5-g]quinazolin-7(6H)-yl)butanoic acid ≥97%
Supplier: Matrix Scientific
115108-1G
115108-500MG
102895-426EA
1040
USD
102895-426
102895-424
4-(6,8-Dioxo-5,8-dihydro[1,3]dioxolo[4,5-g]quinazolin-7(6H)-yl)butanoic acid ≥97%
4-(6,8-Dioxo-5,8-dihydro[1,3]dioxolo[4,5-g]quinazolin-7(6H)-yl)butanoic acid
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₆ |
MDL Number:
MFCD07655292 CAS Number: 892293-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...