6-(3-Methyl-1,2,4-oxadiazol-5-yl)-4,5-dihydropyridazin-3(2H)-one ≥90%

Supplier: Matrix Scientific

115105-500MG 115105-1G
102898-964EA 693.85 USD
102898-964 102898-966
6-(3-Methyl-1,2,4-oxadiazol-5-yl)-4,5-dihydropyridazin-3(2H)-one ≥90%
6-(3-Methyl-1,2,4-oxadiazol-5-yl)-4,5-dihydropyridazin-3(2H)-one

Hazard: Irritant

Formula: C₇H₈N₄O₂
MDL Number: MFCD18916533
CAS Number: 1338682-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR