4-Chloro-2-(3-fluorophenyl)pyrazolo[1,5-a]pyrazine ≥97%

Supplier: Matrix Scientific

115086-500MG 115086-1G
102895-390EA 693.85 USD
102895-390 102895-392
4-Chloro-2-(3-fluorophenyl)pyrazolo[1,5-a]pyrazine ≥97%
4-Chloro-2-(3-fluorophenyl)pyrazolo[1,5-a]pyrazine

Hazard: Irritant

Formula: C₁₂H₇ClFN₃
MDL Number: MFCD25951926
CAS Number: 1338690-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR