Ethyl-4-amino-3-(2-phenylethyl)-2-thioxo-2,3-dihydro-1,3-thiazole-5-carboxylate ≥97%
Supplier: Matrix Scientific
115083-1G
115083-500MG
102895-380EA
1040
USD
102895-380
102895-378
Ethyl-4-amino-3-(2-phenylethyl)-2-thioxo-2,3-dihydro-1,3-thiazole-5-carboxylate ≥97%
Ethyl-4-amino-3-(2-phenylethyl)-2-thioxo-2,3-dihydro-1,3-thiazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₆N₂O₂S₂ MW: 308.42 g/mol |
MDL Number:
MFCD02089350 CAS Number: 312922-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...