3-[1-(3-Fluorophenyl)-1H-1,2,3-triazol-4-yl]-1,2,4-oxadiazole-5-carbohydrazide ≥97%

Supplier: Matrix Scientific

115063-1G 115063-500MG
102895-328EA 693.85 USD
102895-328 102891-420
3-[1-(3-Fluorophenyl)-1H-1,2,3-triazol-4-yl]-1,2,4-oxadiazole-5-carbohydrazide ≥97%
3-[1-(3-Fluorophenyl)-1H-1,2,3-triazol-4-yl]-1,2,4-oxadiazole-5-carbohydrazide

Hazard: Irritant

Formula: C₁₁H₈FN₇O₂
MDL Number: MFCD17780208
CAS Number: 1255783-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR