1-Ethyl-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one ≥97%

Supplier: Matrix Scientific

115055-500MG 115055-1G
102891-274EA 1040 USD
102891-274 102891-272
1-Ethyl-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one ≥97%
1-Ethyl-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one

Hazard: Irritant

Formula: C₉H₁₀N₂O
MDL Number: MFCD20441705
CAS Number: 1340902-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR