3-Amino-4-methoxybut-2-enenitrile ≥97%
Supplier: Matrix Scientific
115018-1G
102895-208EA
958.46
USD
102895-208
3-Amino-4-methoxybut-2-enenitrile ≥97%
3-Amino-4-methoxybut-2-enenitrile
Hazard: Irritant
Formula:
C₅H₈N₂O |
MDL Number:
MFCD19214290 CAS Number: 168686-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...