2-Methoxyethanimidamide ≥97%

Supplier: Matrix Scientific

115014-1G
102895-200EA 738.46 USD
102895-200
2-Methoxyethanimidamide ≥97%
2-Methoxyethanimidamide

Hazard: Irritant

Formula: C₃H₈N₂O
MDL Number: MFCD05663316
CAS Number: 3122-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR