Ethyl-2-methoxyethanimidoate hydrochloride ≥97%

Supplier: Matrix Scientific

115012-1G
102895-196EA 1112.31 USD
102895-196
Ethyl-2-methoxyethanimidoate hydrochloride ≥97%
Ethyl-2-methoxyethanimidoate hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₂
MDL Number: MFCD25963422
CAS Number: 42945-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR