N-(2,2-Dimethyl-4-oxochroman-6-yl)acetamide ≥97%

Supplier: Matrix Scientific

115006-1G
102895-182EA 1126.15 USD
102895-182
N-(2,2-Dimethyl-4-oxochroman-6-yl)acetamide ≥97%
N-(2,2-Dimethyl-4-oxochroman-6-yl)acetamide

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD19380257
CAS Number: 186774-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR