1H-Pyrrolo[2,3-b]pyridine-3-carboxamide ≥97%

Supplier: Matrix Scientific

115005-1G
102895-180EA 958.46 USD
102895-180
1H-Pyrrolo[2,3-b]pyridine-3-carboxamide ≥97%
1H-Pyrrolo[2,3-b]pyridine-3-carboxamide

Hazard: Irritant

Formula: C₈H₇N₃O
MDL Number: MFCD11054202
CAS Number: 74420-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR