(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone ≥97%

Supplier: Matrix Scientific

115002-1G
102895-174EA 958.46 USD
102895-174
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone ≥97%
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone

Hazard: Irritant

Formula: C₁₃H₈ClN₃O
MDL Number: MFCD00108116
CAS Number: 90734-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR