3-Cyclopentylpropionamide ≥95%

Supplier: Matrix Scientific

114994-1G
102895-160EA 787.69 USD
102895-160
3-Cyclopentylpropionamide ≥95%
3-Cyclopentylpropionamide

Hazard: Irritant

Formula: C₈H₁₅NO
Melting Pt: 119…121 °C
MDL Number: MFCD04070174
CAS Number: 935-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR