1-(4-Bromopyrazol-1-yl)-2-chloroethanone ≥97%

Supplier: Matrix Scientific

114978-1G
102899-044EA 958.46 USD
102899-044
1-(4-Bromopyrazol-1-yl)-2-chloroethanone ≥97%
1-(4-Bromopyrazol-1-yl)-2-chloroethanone

Hazard: Irritant

Formula: C₅H₄BrClN₂O
MDL Number: MFCD03419415
CAS Number: 514217-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR