3-Methoxythiophene-2-carboxamide ≥97%

Supplier: Matrix Scientific

114963-1G
102895-102EA 813.85 USD
102895-102
3-Methoxythiophene-2-carboxamide ≥97%
3-Methoxythiophene-2-carboxamide

Hazard: Irritant

Formula: C₆H₇NO₂S
MDL Number: MFCD01680278
CAS Number: 78031-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR