3-Methylpyridine-2-carboxamidine ≥97%

Supplier: Matrix Scientific

114958-1G
102895-090EA 1298.46 USD
102895-090
3-Methylpyridine-2-carboxamidine ≥97%
3-Methylpyridine-2-carboxamidine

Hazard: Irritant

Formula: C₇H₉N₃
MDL Number: MFCD09751910
CAS Number: 742028-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR