2-Acetyl-6-methylpyridine ≥97%
Supplier: Matrix Scientific
114943-1G
102895-028EA
156.92
USD
102895-028
2-Acetyl-6-methylpyridine ≥97%
2-Acetyl-6-methylpyridine
Hazard: Irritant
Formula:
C₈H₉NO MW: 135.17 g/mol |
MDL Number:
MFCD06658292 CAS Number: 6940-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...