1-(6-Methylpyridin-2-yl)propan-1-one ≥97%

Supplier: Matrix Scientific

114939-1G
102895-020EA 813.85 USD
102895-020
1-(6-Methylpyridin-2-yl)propan-1-one ≥97%
1-(6-Methylpyridin-2-yl)propan-1-one

Hazard: Irritant

Formula: C₉H₁₁NO
MDL Number: MFCD18363963
CAS Number: 165126-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR