1-Chloro-5-methoxy-4-nitro-2-(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

114932-1G
102895-006EA 1443.08 USD
102895-006
1-Chloro-5-methoxy-4-nitro-2-(trifluoromethyl)benzene ≥97%
1-Chloro-5-methoxy-4-nitro-2-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₅ClF₃NO₃
MDL Number: MFCD00119612
CAS Number: 646989-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR