2-Chloro-4-methoxy-1-(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

114931-1G
102895-004EA 958.46 USD
102895-004
2-Chloro-4-methoxy-1-(trifluoromethyl)benzene ≥97%
2-Chloro-4-methoxy-1-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₆ClF₃O
MDL Number: MFCD13185465
CAS Number: 1214346-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR