2-Azido-1,3-difluorobenzene ≥97%

Supplier: Matrix Scientific

114927-1G
102894-996EA 1175.38 USD
102894-996
2-Azido-1,3-difluorobenzene ≥97%
2-Azido-1,3-difluorobenzene

Hazard: Irritant

Formula: C₆H₃F₂N₃
MDL Number: MFCD11207349
CAS Number: 102284-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR