1,1,1-Trifluoro-3-phenylpropan-2-ol ≥97%

Supplier: Matrix Scientific

114922-1G
102894-984EA 738.46 USD
102894-984
1,1,1-Trifluoro-3-phenylpropan-2-ol ≥97%
1,1,1-Trifluoro-3-phenylpropan-2-ol

Hazard: Irritant

Formula: C₉H₉F₃O
MDL Number: MFCD00178722
CAS Number: 330-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR