2-[(E)-3-(Dimethylamino)-1-phenylprop-2-enylidene]propanedinitrile ≥97%

Supplier: Matrix Scientific

114912-1G
102894-960EA 958.46 USD
102894-960
2-[(E)-3-(Dimethylamino)-1-phenylprop-2-enylidene]propanedinitrile ≥97%
2-[(E)-3-(Dimethylamino)-1-phenylprop-2-enylidene]propanedinitrile

Hazard: Irritant

Formula: C₁₄H₁₃N₃
MDL Number: MFCD00829180
CAS Number: 124571-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR