4-Methylbenzenesulfinamide ≥97%

Supplier: Matrix Scientific

114911-1G
102894-958EA 813.85 USD
102894-958
4-Methylbenzenesulfinamide ≥97%
4-Methylbenzenesulfinamide

Hazard: Irritant

Formula: C₇H₉NOS
MDL Number: MFCD02093698
CAS Number: 6873-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR