N-Methoxy-4-methylbenzamide ≥97%

Supplier: Matrix Scientific

114909-1G
102894-954EA 1126.15 USD
102894-954
N-Methoxy-4-methylbenzamide ≥97%
N-Methoxy-4-methylbenzamide

Hazard: Irritant

Formula: C₉H₁₁NO₂
MDL Number: MFCD21100905
CAS Number: 25563-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR