2-(2-Fluorophenyl)acetamide ≥97%

Supplier: Matrix Scientific

114899-1G
102894-934EA 716.92 USD
102894-934
2-(2-Fluorophenyl)acetamide ≥97%
2-(2-Fluorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₈FNO
MDL Number: MFCD11643209
CAS Number: 326-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR