2-Fluoro-6-iodobenzamide ≥97%

Supplier: Matrix Scientific

114898-1G
102894-932EA 958.46 USD
102894-932
2-Fluoro-6-iodobenzamide ≥97%
2-Fluoro-6-iodobenzamide

Hazard: Irritant

Formula: C₇H₅FINO
MDL Number: MFCD18396919
CAS Number: 676445-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR