3,5-Dichloro-4-hydroxybenzamide ≥97%

Supplier: Matrix Scientific

114897-1G
102894-930EA 1046.15 USD
102894-930
3,5-Dichloro-4-hydroxybenzamide ≥97%
3,5-Dichloro-4-hydroxybenzamide

Hazard: Irritant

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD06449734
CAS Number: 3337-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR