2-Fluoro-4-methoxybenzamide ≥97%

Supplier: Matrix Scientific

114895-1G
102894-926EA 180.55 USD
102894-926
2-Fluoro-4-methoxybenzamide ≥97%
2-Fluoro-4-methoxybenzamide

Hazard: Irritant

Formula: C₈H₈FNO₂
MDL Number: MFCD01117440
CAS Number: 874804-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR