N-(2,5-Dimethylphenyl)acetamide ≥97%

Supplier: Matrix Scientific

114882-1G
102894-898EA 813.85 USD
102894-898
N-(2,5-Dimethylphenyl)acetamide ≥97%
N-(2,5-Dimethylphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₃NO
MDL Number: MFCD00220610
CAS Number: 2050-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR