N-(2-Acetylphenyl)formamide ≥97%

Supplier: Matrix Scientific

114875-1G
102894-866EA 738.46 USD
102894-866
N-(2-Acetylphenyl)formamide ≥97%
N-(2-Acetylphenyl)formamide

Hazard: Irritant

Formula: C₉H₉NO₂
Melting Pt: 78 °C
MDL Number: MFCD02090023
CAS Number: 5257-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR