N1-(4-Nitrophenyl)-2,4-dinitroaniline ≥97%

Supplier: Matrix Scientific

114874-1G
102894-864EA 738.46 USD
102894-864
N1-(4-Nitrophenyl)-2,4-dinitroaniline ≥97%
N1-(4-Nitrophenyl)-2,4-dinitroaniline

Hazard: Irritant

Formula: C₁₂H₈N₄O₆
MDL Number: MFCD00099134
CAS Number: 970-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR