2,5-Dimethyl-p-anisonitrile ≥97%

Supplier: Matrix Scientific

114872-1G
102894-860EA 253.85 USD
102894-860
2,5-Dimethyl-p-anisonitrile ≥97%
2,5-Dimethyl-p-anisonitrile

Hazard: Irritant

Formula: C₁₀H₁₁NO
MDL Number: MFCD00053416
CAS Number: 64400-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR