4-(2-Fluorophenyl)but-3-en-2-one ≥97%

Supplier: Matrix Scientific

114871-1G
102894-858EA 400 USD
102894-858
4-(2-Fluorophenyl)but-3-en-2-one ≥97%
4-(2-Fluorophenyl)but-3-en-2-one

Hazard: Irritant

Formula: C₁₀H₉FO
MW: 164.18 g/mol
MDL Number: MFCD01221416
CAS Number: 2143-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR