1-(3,4-Chlorophenyl)-2-methoxyethanone ≥97%

Supplier: Matrix Scientific

114869-1G
102894-854EA 641.54 USD
102894-854
1-(3,4-Chlorophenyl)-2-methoxyethanone ≥97%
1-(3,4-Chlorophenyl)-2-methoxyethanone

Hazard: Irritant

Formula: C₉H₈Cl₂O₂
MDL Number: MFCD11555895
CAS Number: 1157136-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR