tert-Butyl-N-[4-(cyanomethyl)phenyl]carbamate ≥97%

Supplier: Matrix Scientific

114866-1G 114866-10G
102894-846EA 364.62 USD
102894-846 102894-848
tert-Butyl-N-[4-(cyanomethyl)phenyl]carbamate ≥97%
tert-Butyl-N-[4-(cyanomethyl)phenyl]carbamate

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
MDL Number: MFCD17465121
CAS Number: 1233249-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR