3-Chloro-1-(4-methoxyphenyl)prop-2-en-1-one ≥97%

Supplier: Matrix Scientific

114858-1G
102894-828EA 813.85 USD
102894-828
3-Chloro-1-(4-methoxyphenyl)prop-2-en-1-one ≥97%
3-Chloro-1-(4-methoxyphenyl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₀H₉ClO₂
MDL Number: MFCD01673992
CAS Number: 15724-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR