4-(Bromomethyl)-1-methyl-1H-pyrazole hydrobromide ≥97%

Supplier: Matrix Scientific

114850-1G
102899-030EA 1298.46 USD
102899-030
4-(Bromomethyl)-1-methyl-1H-pyrazole hydrobromide ≥97%
4-(Bromomethyl)-1-methyl-1H-pyrazole hydrobromide

Hazard: Irritant

Formula: C₅H₈Br₂N₂
MW: 255.95 g/mol
MDL Number: MFCD13195912
CAS Number: 528878-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR