3-Bromo-4-(bromomethyl)thiophene ≥95%
Supplier: Matrix Scientific
114846-1G
102898-926EA
932.31
USD
102898-926
3-Bromo-4-(bromomethyl)thiophene ≥95%
3-Bromo-4-(bromomethyl)thiophene
Hazard: Irritant
Formula:
C₅H₄Br₂S |
MDL Number:
MFCD09817482 CAS Number: 40032-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...