2-(1H-Tetrazol-1-yl)acetamide ≥97%

Supplier: Matrix Scientific

114840-1G
102894-814EA 910.77 USD
102894-814
2-(1H-Tetrazol-1-yl)acetamide ≥97%
2-(1H-Tetrazol-1-yl)acetamide

Hazard: Irritant

Formula: C₃H₅N₅O
MDL Number: MFCD14525648
CAS Number: 26476-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR