2-Methylpyrrolidine ≥97%

Supplier: Matrix Scientific

114836-10G 114836-1G
102894-806EA 1090.77 USD
102894-806 102894-804
2-Methylpyrrolidine ≥97%
2-Methylpyrrolidine

Hazard: Irritant

Formula: C₅H₁₁N
MW: 85.15 g/mol
MDL Number: MFCD00014491
CAS Number: 765-38-8
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR