Quinoline-2-carboxamide ≥97%

Supplier: Matrix Scientific

114827-1G
102894-786EA 910.77 USD
102894-786
Quinoline-2-carboxamide ≥97%
Quinoline-2-carboxamide

Hazard: Irritant

Formula: C₁₀H₈N₂O
MDL Number: MFCD01366548
CAS Number: 5382-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR