Pimobendan ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(2-Methyl-1-oxopropyl)isoindole-1,3-dione

114825-1G
102894-782EA 1104.62 USD
102894-782
Pimobendan ≥97%
Pimobendan

Hazard: Irritant

Formula: C₁₉H₁₈N₄O₂
MW: 334.38 g/mol
MDL Number: MFCD00761648
CAS Number: 74150-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR