5-Acetyl-2,3-dihydro-1H-indol-2-one ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Acetyloxindole

114818-1G
102894-768EA 641.54 USD
102894-768
5-Acetyl-2,3-dihydro-1H-indol-2-one ≥97%
5-Acetyl-2,3-dihydro-1H-indol-2-one

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD07782089
CAS Number: 64483-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR