6-Oxo-1,6-dihydropyridazine-3-carboxamide ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxypyridazine-3-carboxamide

114814-1G
102894-758EA 813.85 USD
102894-758
6-Oxo-1,6-dihydropyridazine-3-carboxamide ≥97%
6-Oxo-1,6-dihydropyridazine-3-carboxamide

Hazard: Irritant

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
MDL Number: MFCD11643204
CAS Number: 60184-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR