4-(Trifluoromethyl)pyrimidine ≥97%

Supplier: Matrix Scientific

114812-1G
102894-752EA 438.46 USD
102894-752
4-(Trifluoromethyl)pyrimidine ≥97%
4-(Trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₃F₃N₂
MDL Number: MFCD13186003
CAS Number: 136547-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR