2-(Methoxymethyl)pyrimidine-4,6-diol ≥97%

Supplier: Matrix Scientific

114808-1G
102894-744EA 813.85 USD
102894-744
2-(Methoxymethyl)pyrimidine-4,6-diol ≥97%
2-(Methoxymethyl)pyrimidine-4,6-diol

Hazard: Irritant

Formula: C₆H₈N₂O₃
MDL Number: MFCD21337360
CAS Number: 1903-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR