Pyrimidine-4-carboxamide ≥97%

Supplier: Matrix Scientific

114807-1G
102894-742EA 835.38 USD
102894-742
Pyrimidine-4-carboxamide ≥97%
Pyrimidine-4-carboxamide

Hazard: Irritant

Formula: C₅H₅N₃O
MDL Number: MFCD17013173
CAS Number: 28648-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR