6-(Bromomethyl)pyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Bromomethyl-2-cyanopyridine6-(Bromomethyl)picolinonitrile

114806-1G
102894-740EA 958.46 USD
102894-740
6-(Bromomethyl)pyridine-2-carbonitrile ≥97%
6-(Bromomethyl)pyridine-2-carbonitrile

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
MDL Number: MFCD10687145
CAS Number: 104508-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR